3-(Methylsulfonyl)benzoic acid ≥98%

Supplier: BeanTown Chemical

125450-1G 125450-5G
BT125450-1GEA 66.82 USD
BT125450-1G BT125450-5G
3-(Methylsulfonyl)benzoic acid ≥98%
3-(Methylsulphonyl)benzoic acid

CAS: 5345-27-7; MDL No: MFCD00216488 Powder; Molecular Formula: C8H8O4S; MW: 200.21 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₈H₈O₄S
MW: 200.21 g/mol
MDL Number: MFCD00216488
CAS Number: 5345-27-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR