3-Amino-2,2-dimethyl-1-propanol ≥95%

Supplier: BeanTown Chemical

125435-1G 125435-5G
BT125435-1GEA 32 USD
BT125435-1G BT125435-5G
3-Amino-2,2-dimethyl-1-propanol ≥95%
3-Amino-2,2-dimethyl-1-propanol

CAS: 26734-09-8; EC No: 247-948-3; MDL No: MFCD00059847 Solid; Molecular Formula: C5H13NO; MW: 103.17 Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₅H₁₃NO
MW: 103.17 g/mol
MDL Number: MFCD00059847
CAS Number: 26734-09-8
EINECS: 247-948-3

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR