4-Iodo-3-nitrobenzoic acid ≥97%
Supplier: BeanTown Chemical
125420-25G
125420-5G
BT125420-25GEA
721
USD
BT125420-25G
BT125420-5G
4-Iodo-3-nitrobenzoic acid ≥97%
4-Iodo-3-nitrobenzoic acid
CAS: 35674-27-2; MDL No: MFCD00502399
Crystalline/Powder; Molecular Formula: C7H4INO4; MW: 293.02
Light Sensitive
- Light Sensitive
- Crystalline/Powder
Formula:
C₇H₄INO₄ MW: 293.02 g/mol |
MDL Number:
MFCD00502399 CAS Number: 35674-27-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...