Ethyl-2-phenyl-2-cyanobutylate

Supplier: BeanTown Chemical

Synonyms: Ethyl 2-cyano-2-phenylbutanoate

125395-1G 125395-5G
BT125395-1GEA 74 USD
BT125395-1G BT125395-5G
Ethyl-2-phenyl-2-cyanobutylate
Ethyl-2-phenyl-2-cyanobutylate

CAS: 718-71-8; MDL No: MFCD00068977 Liquid; Molecular Formula: C13H15NO2; MW: 217.27


  • Liquid

Formula: C₁₃H₁₅NO₂
MW: 217.268 g/mol
MDL Number: MFCD00068977
CAS Number: 718-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR