1-Hexanesulfonic acid sodium salt ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium hexane-1-sulphonate, Sodium hexanesulphonate, Sodium 1-hexanesulphonate
125345-100G
125345-25G
125345-5G
BT125345-100GEA
529.99
USD
BT125345-100G
BT125345-25G
BT125345-5G
1-Hexanesulfonic acid sodium salt ≥99%
1-Hexanesulphonic acid sodium salt
CAS: 2832-45-3; EC No: 220-601-3; MDL No: MFCD00007542
Solid; Linear Formula: CH3(CH2)5SO3Na; MW: 188.22
Melting Point:
- Solid
Formula:
C₆H₁₃O₃SNa MW: 188.22 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00007542 CAS Number: 2832-45-3 EINECS: 220-601-3 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...