4,5-Dibromo-3(2H)-pyridazinone ≥98%
Supplier: BeanTown Chemical
125325-25G
125325-5G
BT125325-25GEA
443
USD
BT125325-25G
BT125325-5G
4,5-Dibromo-3(2H)-pyridazinone ≥98%
4,5-Dibromo-3(2H)-pyridazinone
CAS: 5788-58-9; MDL No: MFCD00023641
Solid; Molecular Formula: C4H2Br2N2O; MW: 253.88
Melting Point: 220° (decomposes)
- Solid
Formula:
C₄H₂Br₂N₂O Melting Pt: 220 °C (dec.) |
MDL Number:
MFCD00023641 CAS Number: 5788-58-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...