4-Perhydroazepinone hydrochloride ≥96%

Supplier: BeanTown Chemical

Synonyms: 4-Azepanone hydrochloride, Hexahydro-4-azepinone hydrochloride

125300-1G 125300-25G 125300-5G
BT125300-1GEA 42 USD
BT125300-1G BT125300-25G BT125300-5G
4-Perhydroazepinone hydrochloride ≥96%
4-Perhydroazepinone hydrochloride

CAS: 50492-22-3; MDL No: MFCD02183487 Solid; Molecular Formula: C6H11NO·HCl; MW: 149.62 Melting Point: 176-181°; Flash point: 152°C (306°F)


  • Solid

Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
Melting Pt: 176…181 °C
Flash Pt: 152 °C (306 °F)
MDL Number: MFCD02183487
CAS Number: 50492-22-3

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR