4-Phenylimidazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Phenyl-1H-imidazole
125285-25G
125285-5G
BT125285-25GEA
312
USD
BT125285-25G
BT125285-5G
4-Phenylimidazole ≥98%
4-Phenylimidazole
CAS: 670-95-1; EC No: 211-580-1; MDL No: MFCD00005197
Granular; Molecular Formula: C9H8N2; MW: 144.17
Melting Point: 128-131°
- Granular
Formula:
C₉H₈N₂ MW: 144.18 g/mol Melting Pt: 128…131 °C |
MDL Number:
MFCD00005197 CAS Number: 670-95-1 EINECS: 211-580-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...