Bis(pinacolato)diboron ≥98%

Supplier: BeanTown Chemical

125265-1G 125265-25G 125265-5G
BT125265-1GEA 39 USD
BT125265-1G BT125265-25G BT125265-5G
Bis(pinacolato)diboron ≥98%
Formula: C₁₂H₂₄B₂O₄
Melting Pt: 137…140 °C
MDL Number: MFCD00799570
CAS Number: 73183-34-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR