4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide

Supplier: BeanTown Chemical

Synonyms: 2,6,7-Trioxa-1-phosphabicyclo[2.2.2]octane-4-methanol 1-oxide

125175-25G 125175-5G
BT125175-25GEA 352 USD
BT125175-25G BT125175-5G
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide

CAS: 5301-78-0; RTECS: YK0690700 Powder; Molecular Formula: C5H9O5P; MW: 180.10


  • RTECS: YK0690700
  • Powder

Formula: C₅H₉O₅P
MW: 180.096 g/mol
MDL Number: MFCD01677612
CAS Number: 5301-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR