Diosmin ≥95%
Supplier: BeanTown Chemical
Synonyms:
5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
125165-100G
125165-25G
125165-5G
BT125165-100GEA
371
USD
BT125165-100G
BT125165-25G
BT125165-5G
Diosmin ≥95%
Diosmin
CAS: 520-27-4; EC No: 208-289-7; MDL No: MFCD00009772
Solid; Molecular Formula: C28H32O15; MW: 608.54
Formula:
C₂₈H₃₂O₁₅ MW: 608.5 g/mol |
MDL Number:
MFCD00009772 CAS Number: 520-27-4 EINECS: 208-289-7 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...