Palladium(II) acetate trimer

Supplier: BeanTown Chemical

Synonyms: Palladium diacetate, Palladium (II) acetate

125140-1G 125140-5G
BT125140-1GEA 142.61 USD
BT125140-1G BT125140-25G BT125140-5G
Palladium(II) acetate trimer
Palladium(II) acetate

CAS: 3375-31-3; EC No: 222-164-4; MDL No: MFCD00012453; RTECS: AJ9000000 Needles; Linear Formula: [Pd(OOCCH3)2]3; MW: 673.46 Melting Point: 205° (decomposes)


  • RTECS: AJ9000000
  • Needles

Formula: C₄H₆O₄Pd
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR