Methyl-4-iodo-3-methylbenzoate ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 4-iodo-3-methylbenzoate

125120-100G 125120-25G 125120-5G
BT125120-100GEA 461 USD
BT125120-100G BT125120-25G BT125120-5G
Methyl-4-iodo-3-methylbenzoate ≥98%
Methyl 4-iodo-3-methylbenzoate

CAS: 5471-81-8; MDL No: MFCD02683863 Crystalline; Molecular Formula: C9H9IO2; MW: 276.07 Melting Point: 59-62° Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: C₉H₉IO₂
MW: 276.07 g/mol
Melting Pt: 59…62 °C
MDL Number: MFCD02683863
CAS Number: 5471-81-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR