Homovanillic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-(4-Hydroxy-3-methoxyphenyl)acetic acid, (4-Hydroxy-3-methoxyphenyl)acetic acid
125105-1G
125105-25G
125105-5G
BT125105-1GEA
60
USD
BT125105-1G
BT125105-25G
BT125105-5G
Homovanillic acid ≥98%
Homovanillic acid
CAS: 306-08-1; EC No: 206-176-7; MDL No: MFCD00004350
Powder; Molecular Formula: C9H10O4; MW: 182.18
Melting Point: 142-145°
- Powder
Formula:
(C₆H₃(OH)(OCH₃))CH₂COOH MW: 182.18 g/mol Melting Pt: 142…145 °C |
MDL Number:
MFCD00004350 CAS Number: 306-08-1 EINECS: 206-176-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...