Poly(acrylic acid) solution, M.W. ≈ 240,000 25% (w/w) in water

Supplier: BeanTown Chemical

125080-100G 125080-500G
BT125080-100GEA 58.24 USD
BT125080-100G BT125080-500G
Poly(acrylic acid) solution, M.W. ≈ 240,000 25% (w/w) in water
Poly(acrylic acid)

CAS: 9003-01-4; MDL No: MFCD00084394 Liquid; Linear Formula: [-CH2CH(CO2H)-]n; MW ≈240,000 Density (g/mL): 1.15


  • Liquid

Formula: C₃H₄O₂
Density: 1.15 g/mL
MDL Number: MFCD00084394
CAS Number: 9003-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR