Vanillin ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

125060-100G 125060-2.5KG 125060-500G
BT125060-100GEA 34.03 USD
BT125060-100G BT125060-2.5KG BT125060-500G
Vanillin ≥99%
Vanillin

CAS: 121-33-5; EC No: 204-465-2; MDL No: MFCD00006942; RTECS: YW5775000 Solid; Linear Formula: (HO)C6H3(OCH3)CHO; Molecular Formula: C8H8O3; MW: 152.15 Melting Point: 81-83°; Boiling Point: 170°/15 mmHg; Flash point: 153°C (307°F) Density (g/mL): 1.056 Air Sensitive, Light Sensitive, Moisture Sensitive


  • RTECS: YW5775000
  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Solid

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 170 °C/15 mmHg
Melting Pt: 81…83 °C
Density: 1.056 g/mL
Flash Pt: 153 °C (307 °F)
MDL Number: MFCD00006942
CAS Number: 121-33-5
EINECS: 204-465-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR