1H-Pyrazole-4-boronic acid pinacol ester ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

125050-1G 125050-5G
BT125050-1GEA 91 USD
BT125050-1G BT125050-5G
1H-Pyrazole-4-boronic acid pinacol ester ≥98%
1H-Pyrazole-4-boronic acid pinacol ester

CAS: 269410-08-4; MDL No: MFCD03453063 Powder; Molecular Formula: C9H15BN2O2; MW: 194.04 Melting Point: 142-146°


  • Powder

Formula: C₉H₁₅BN₂O₂
Melting Pt: 142…146 °C
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR