3-Amino-1-bromoisoquinoline ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-Bromoisoquinolin-3-amine
125025-1G
125025-5G
BT125025-1GEA
203
USD
BT125025-1G
BT125025-5G
3-Amino-1-bromoisoquinoline ≥97%
3-Amino-1-bromoisoquinoline
CAS: 13130-79-5; MDL No: MFCD00102193
Solid; Molecular Formula: C9H7BrN2; MW: 223.07
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₉H₇BrN₂ MW: 223.07 g/mol |
MDL Number:
MFCD00102193 CAS Number: 13130-79-5 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...