Titanium diboride ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
125010-10G
125010-250G
125010-50G
BT125010-10GEA
40
USD
BT125010-10G
BT125010-250G
BT125010-50G
Titanium diboride ≥99.5% (trace metals basis)
Titanium diboride
CAS: 12045-63-5; EC No: 234-961-4; MDL No: MFCD00049595
-325 Mesh Powder; Linear Formula: TiB2; MW: 69.50
Melting Point: 3225°
Density (g/mL): 4.52
- -325 Mesh Powder
Formula:
B₂Ti Melting Pt: 3225 °C Density: 4.52 g/mL |
MDL Number:
MFCD00049595 CAS Number: 12045-63-5 EINECS: 234-961-4 |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...