Ammonium carbonate ≥30% (NH₃ basis) ACS

Supplier: BeanTown Chemical

Synonyms: Salt of hartshorn

125005-2KG 125005-500G
BT125005-2KGEA 133 USD
BT125005-2KG BT125005-500G
Ammonium carbonate ≥30% (NH₃ basis) ACS
Ammonium carbonate

CAS: 506-87-6; EC No: 208-058-0; MDL No: MFCD00010890; RTECS: BP1925000 Powder; Linear Formula: (NH4)2CO3; MW: 96.09 Hygroscopic


  • RTECS: BP1925000
  • Hygroscopic
  • Powder

Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR