myo-Inositol ≥98%

Supplier: BeanTown Chemical

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

124990-100G 124990-2.5KG 124990-500G
BT124990-100GEA 57.97 USD
BT124990-100G BT124990-2.5KG BT124990-500G
myo-Inositol ≥98%
myo-Inositol

CAS: 87-89-8; EC No: 201-781-2; MDL No: MFCD00077932; RTECS: NM7520800 Crystalline/Powder; Molecular Formula: C6H12O6; MW: 180.16 Melting Point: 222-227° Density (g/mL): 1.75


  • RTECS: NM7520800
  • Crystalline/Powder

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 222…227 °C
Density: 1.75 g/mL
MDL Number: MFCD00077932
CAS Number: 87-89-8
EINECS: 201-781-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR