2,4,6-Triisopropylphenylboronic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,4,6-Triisopropylbenzeneboronic acid, (2,4,6-Triisopropylphenyl)boronic acid
124960-1G
124960-25G
124960-5G
BT124960-1GEA
112
USD
BT124960-1G
BT124960-25G
BT124960-5G
2,4,6-Triisopropylphenylboronic acid ≥98%
2,4,6-Triisopropylphenylboronic acid
CAS: 154549-38-9; MDL No: MFCD02683099
Solid; Molecular Formula: C15H25BO2; MW: 248.16
Melting Point: 167-172°
- Solid
Formula:
C₁₅H₂₅BO₂ Melting Pt: 167…172 °C |
MDL Number:
MFCD02683099 CAS Number: 154549-38-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...