Bismuth(III) oxide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
124940-250G
124940-50G
BT124940-250GEA
253
USD
BT124940-250G
BT124940-50G
Bismuth(III) oxide ≥99.99%
Bismuth(III) oxide
CAS: 1304-76-3; EC No: 215-134-7; MDL No: MFCD00003462; RTECS: EB2984460
-325 Mesh Powder ; Molecular Formula: Bi2O3; MW: 465.96
- RTECS: EB2984460
- -325 Mesh Powder
Formula:
BiO₃ MW: 465.96 g/mol Boiling Pt: 1890 Melting Pt: 825 |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 13,01273 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...