Acetonylacetone ≥97%

Supplier: BeanTown Chemical

Synonyms: Hexane-2,5-dione, 2,5-Hexanedione

124935-100G 124935-25G 124935-500G
BT124935-100GEA 55 USD
BT124935-100G BT124935-25G BT124935-500G
Acetonylacetone ≥97%
Acetonylacetone

CAS: 110-13-4; EC No: 203-738-3; MDL No: MFCD00008792; RTECS: MO3150000 Liquid; Molecular Formula: C6H10O2; MW: 114.14 Melting Point: -6°; Boiling Point: 191°; Flash point: 79°C (174°F) Density (g/mL): 0.973; Refractive Index: 1.425


  • RTECS: MO3150000
  • Refractive index: 1.425
  • Liquid

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 191 °C
Melting Pt: –6 °C
Density: 0.973 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00008792
CAS Number: 110-13-4
EINECS: 203-738-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR