2-(2-Ethoxyethoxy)ethyl acetate ≥99%

Supplier: BeanTown Chemical

Synonyms: Diethylene glycol monoethyl ether acetate

124915-100ML 124915-2.5L 124915-500ML
BT124915-100MLEA 24 USD
BT124915-100ML BT124915-2.5L BT124915-500ML
2-(2-Ethoxyethoxy)ethyl acetate ≥99%
2-(2-Ethoxyethoxy)ethyl acetate

CAS: 112-15-2; EC No: 203-940-1; MDL No: MFCD00041928; RTECS: KK8925000 Liquid; Linear Formula: CH3CH2OCH2CH2OCH2CH2CO2CH3; Molecular Formula: C8H16O4; MW: 176.21 Boiling Point: 218-219°; Flash point: 98°C (208°F) Density (g/mL): 1.011; Refractive Index: 1.421


  • RTECS: KK8925000
  • Refractive index: 1.421
  • Liquid

Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 218…219 °C
Density: 1.011 g/mL
Flash Pt: 98 °C (208 °F)
MDL Number: MFCD00041928
CAS Number: 112-15-2
EINECS: 203-940-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR