15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) ≥98%
Supplier: BeanTown Chemical
CAS: 33100-27-5; EC No: 251-379-6; MDL No: MFCD00005110; RTECS: SB0200000
Liquid; Molecular Formula: C10H20O5; MW: 220.26
Boiling Point: 93-96°/0.05 mmHg; Flash point:
- RTECS: SB0200000
- Refractive index: 1.465
- Hygroscopic
- Liquid
Formula:
C₁₀H₂₀O₅ Boiling Pt: 93…96 °C/0.05 mmHg Density: 1.113 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00005110 CAS Number: 33100-27-5 EINECS: 251-379-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...