4,7-Dihydroxy-1,10-phenanthroline

Supplier: BeanTown Chemical

Synonyms: 1,10-Phenanthroline-4,7-diol

124895-10G 124895-2G 124895-500MG
BT124895-10GEA 556 USD
BT124895-10G BT124895-2G BT124895-500MG
4,7-Dihydroxy-1,10-phenanthroline
4,7-Dihydroxy-1,10-phenanthroline

CAS: 3922-40-5; EC No: 223-493-6; MDL No: MFCD00004975 Solid; Molecular Formula: C12H8N2O2; MW: 212.20 Melting Point: 300° Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₁₂H₈N₂O₂
MW: 212.21 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00004975
CAS Number: 3922-40-5
EINECS: 223-493-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR