Ammonium molybdate (para) hydrate ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Ammonium molybdate hydrate

124875-25G 124875-5G
BT124875-25GEA 170.08 USD
BT124875-25G BT124875-5G
Ammonium molybdate (para) hydrate ≥99.999% (trace metals basis)
Ammonium molybdate (para) hydrate

CAS: 12027-67-7; EC No: 234-722-4; MDL No: MFCD00064633; RTECS: QA4900000 Powder; Linear Formula: (NH4)6Mo7O24·H2O; MW: 1163.79 (anhydrous) Density (g/mL): 2.498


  • RTECS: QA4900000
  • Powder

Formula: H₂₄Mo₇N₆O₂₄·xH₂O
MW: 1163.8 g/mol (anhy)
Density: 2.498 g/mL
MDL Number: MFCD00064633
CAS Number: 12027-67-7
EINECS: 234-722-4

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR