Calcium metasilicate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Calcium silicate
124870-100G
124870-25G
124870-500G
BT124870-100GEA
164
USD
BT124870-100G
BT124870-25G
BT124870-500G
Calcium metasilicate ≥99%
Calcium metasilicate
CAS: 10101-39-0; EC No: 233-250-6; MDL No: MFCD00015979
-200 Mesh Powder; Linear Formula: CaSiO3 ; MW: 116.16
- -200 Mesh Powder
Formula:
CaO₃Si MW: 116.16 g/mol |
MDL Number:
MFCD00015979 CAS Number: 10101-39-0 EINECS: 233-250-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...