Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Cu(TMHD)2, Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)

124850-1G 124850-25G 124850-5G
BT124850-1GEA 50 USD
BT124850-1G BT124850-25G BT124850-5G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99.9% (trace metals basis)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II)

CAS: 14040-05-2; MDL No: MFCD00058920 Powder; Molecular Formula: C22H38CuO4; MW: 430.08 Melting Point: 198° (decomposes)


  • Powder

Formula: C₂₂H₃₈CuO₄
MW: 430.08 g/mol
Melting Pt: 198 °C (dec.)
MDL Number: MFCD00058920
CAS Number: 14040-05-2

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR