2,2'-Dipyridyl disulfide ≥98%

Supplier: BeanTown Chemical

124820-100G 124820-25G 124820-5G
BT124820-100GEA 602 USD
BT124820-100G BT124820-25G BT124820-5G
2,2'-Dipyridyl disulfide ≥98%
2,2'-Dipyridyl disulphide

CAS: 2127-03-9; EC No: 218-343-1; MDL No: MFCD00006287 Powder; Molecular Formula: C10H8N2S2; MW: 220.3 Melting Point: 56-58°


  • Powder

Formula: C₁₀H₈N₂S₂
Melting Pt: 56…58 °C
MDL Number: MFCD00006287
CAS Number: 2127-03-9
EINECS: 218-343-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR