Sodium diethyldithiocarbamate trihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate

124810-100G 124810-500G
BT124810-100GEA 62.92 USD
BT124810-100G BT124810-500G
Sodium diethyldithiocarbamate trihydrate ≥98%
Sodium diethyldithiocarbamate trihydrate

CAS: 20624-25-3; EC No: 205-710-6; MDL No: MFCD00150617; RTECS: EZ6550000 Crystalline; Linear Formula: (CH2CH2)2NCSSNa·3H2O; Molecular Formula: C5H10NNaS·3H2O; MW: 225.31 Melting Point: 95-99° Density (g/mL): 1.100 Hygroscopic


  • RTECS: EZ6550000
  • Hygroscopic
  • Crystalline

Formula: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Melting Pt: 95…99 °C
Density: 1.100 g/mL
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR