Chromium(III) oxide ≥98% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Chromic oxide
124795-2KG
124795-500G
BT124795-2KGEA
109.61
USD
BT124795-2KG
BT124795-500G
Chromium(III) oxide ≥98% (trace metals basis)
Chromium(III) oxide
CAS: 1308-38-9; EC No: 215-160-9; MDL No: MFCD00010949; RTECS: GB6475000
-325 Mesh Powder; Molecular Formula: Cr2O3; MW: 151.99
- RTECS: GB6475000
- -325 Mesh Powder
Formula:
Cr₂O₃ MW: 151.99 g/mol |
MDL Number:
MFCD00010949 CAS Number: 1308-38-9 EINECS: 215-160-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...