Sodium ammonium hydrogen phosphate tetrahydrate
Supplier: BeanTown Chemical
Synonyms:
XSodium ammonium hydrogen phosphate tetrahydrate, Sodium ammonium phosphate, Ammonium sodium phosphate dibasic tetrahydrate, Sodium ammonium phosphate tetrahydrate, Ammonium sodium phosphate tetrahydrate
124790-100G
124790-2.5KG
124790-500G
BT124790-100GEA
24
USD
BT124790-100G
BT124790-2.5KG
BT124790-500G
Sodium ammonium hydrogen phosphate tetrahydrate
Sodium ammonium hydrogen phosphate tetrahydrate
CAS: 7783-13-3; MDL No: MFCD00066970
Crystalline; Linear Formula: NaNH4HPO4·4H2O; MW: 209.07
Melting Point: 79° (decomposes)
Density (g/mL): 1.544
- Crystalline
Formula:
NaNH₄HPO₄·4H₂O MW: 209.07 g/mol Melting Pt: 79 °C (dec.) Density: 1.544 g/mL |
MDL Number:
MFCD00066970 CAS Number: 7783-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...