Cesium iodide ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Cesiumiodide
124750-100G
124750-25G
BT124750-100GEA
311
USD
BT124750-100G
BT124750-25G
Cesium iodide ≥99.999% (trace metals basis)
Cesium iodide
CAS: 7789-17-5; EC No: 232-145-2; MDL No: MFCD00010962; RTECS: FL0350000
Crystalline; Molecular Formula: CsI; MW: 259.81
Melting Point: 626°
Density (g/mL): 4.51
Hygroscopic
- RTECS: FL0350000
- Hygroscopic
- Crystalline
Formula:
CsI MW: 259.81 g/mol Melting Pt: 626 °C Density: 4.51 g/mL |
MDL Number:
MFCD00010962 CAS Number: 7789-17-5 EINECS: 232-145-2 |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...