1-(3-Hydroxypropyl)piperazine ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Piperazinepropanol, 3-Piperazin-1-ylpropanol, 3-(Piperazin-1-yl)propan-1-ol

124745-1G 124745-5G
BT124745-1GEA 83 USD
BT124745-1G BT124745-5G
1-(3-Hydroxypropyl)piperazine ≥98%
1-(3-Hydroxypropyl)piperazine

CAS: 5317-32-8; EC No: 226-176-0; MDL No: MFCD00023132 Solid; Molecular Formula: C7H16N2O ; MW: 144.21 Melting Point: 49-53°; Flash point: 110°C (230°F)


  • Solid

Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
Melting Pt: 49…53 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00023132
CAS Number: 5317-32-8
EINECS: 226-176-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR