4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate, (∼30% H₂O)

Supplier: BeanTown Chemical
Warning

Synonyms: 2,2'-Benzidinedisulphonic Acid, 4,4'-Diamino-[1,1'-biphenyl]-2,2'-disulphonic acid(contains 30% Water at maximum)

124740-100G 124740-25G
BT124740-100GEA 150.31 USD
BT124740-100G BT124740-25G
4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate, (∼30% H₂O)
4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate

CAS: 117-61-3; EC No: 204-200-0; MDL No: MFCD00041885; RTECS: DV5230000 Powder; Molecular Formula: C12H12N2O6S2·xH2O; MW: 344.36 (anhydrous)


  • RTECS: DV5230000
  • Powder

Formula: C₁₂H₁₂N₂O₆S₂·xH₂O
MW: 344.36 g/mol (anhy)
MDL Number: MFCD00041885
CAS Number: 117-61-3
EINECS: 204-200-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR