Lanthanum(III) carbonate hydrate ≥99.9%

Supplier: BeanTown Chemical

124735-100G 124735-25G 124735-500G
BT124735-100GEA 46 USD
BT124735-100G BT124735-25G BT124735-500G
Lanthanum(III) carbonate hydrate ≥99.9%
Lanthanum(III) carbonate hydrate

CAS: 54451-24-0; EC No: 209-599-5; MDL No: MFCD00011069 Powder; Linear Formula: La2(CO3)3·xH2O ; MW: 457.85 (anhydrous) Hygroscopic


  • Hygroscopic
  • Powder

Formula: La₂(CO₃)₃.xH₂O
MW: 457.85 g/mol (anhy)
MDL Number: MFCD00011069
CAS Number: 54451-24-0
EINECS: 209-599-5

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR