(2,4,6-Trifluorophenyl)acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,4,6-Trifluorophenylacetic acid

124685-2.5G 124685-500MG
BT124685-2.5GEA 290 USD
BT124685-2.5G BT124685-500MG
(2,4,6-Trifluorophenyl)acetic acid ≥98%
(2,4,6-Trifluorophenyl)acetic acid

CAS: 209991-63-9; MDL No: MFCD00061218 Crystalline; Molecular Formula: C8H5F3O2; MW: 190.12


  • Crystalline

Formula: (C₆H₂F₃)CH₂COOH
MW: 190.12 g/mol
MDL Number: MFCD00061218
CAS Number: 209991-63-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR