Iron(II) acetate, anhydrous
Supplier: BeanTown Chemical
124680-100G
124680-25G
BT124680-100GEA
584.71
USD
BT124680-100G
BT124680-25G
Iron(II) acetate, anhydrous
Iron(II) acetate
CAS: 3094-87-9; EC No: 221-441-7; MDL No: MFCD00058909
Powder/Lump; Linear Formula: Fe(OOCCH3)2; MW: 173.94
Melting Point: 199-200° (decomposes)
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder/Lump
Formula:
C₄H₆FeO₄ MW: 173.94 g/mol Melting Pt: 199…200 °C (dec.) |
MDL Number:
MFCD00058909 CAS Number: 3094-87-9 EINECS: 221-441-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...