β-(N-Morpholino)ethanesulfonic acid (MES) hydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Morpholineethanesulphonic acid hydrate, MES hydrate, 2-(4-Morpholinyl)ethanesulphonic acid hydrate

124645-250G 124645-500G
BT124645-250GEA 203.74 USD
BT124645-250G BT124645-500G
β-(N-Morpholino)ethanesulfonic acid (MES) hydrate ≥99%
β-(N-Morpholino)ethanesulfonic acid (MES) hydrate

CAS: 4432-31-9; EC No: 224-632-3; MDL No: MFCD00149409; RTECS: QE9104530 Powder; Molecular Formula: C6H13NO4S·xH2O; MW: 195.24 Melting Point: 308° (decomposes)

Formula: C₆H₁₃NO₄S·xH₂O
MW: 195.24 g/mol (anhy)
MDL Number: MFCD00149409
CAS Number: 1266615-59-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR