Palladium ≥99.95% (trace metals basis), granules
Supplier: BeanTown Chemical
124600-1G
124600-5G
BT124600-1GEA
238
USD
BT124600-1G
BT124600-5G
Palladium ≥99.95% (trace metals basis), granules
Palladium
CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167; RTECS: RT3480500
Solid; Molecular Formula: Pd; MW: 106.42
Melting Point: 1554°; Boiling Point: 2970°
Density (g/mL): 12.02
- RTECS: RT3480500
- Solid
Formula:
Pd MW: 106.420 g/mol Boiling Pt: 2970 °C Melting Pt: 1554 °C Density: 12.02 g/mL |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 |
Specification Test Results
Assay | 99.95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...