Sodium diatrizoate hydrate ≥98%
Supplier: BeanTown Chemical
|
Danger
|
124590-10G
124590-250G
124590-50G
BT124590-10GEA
25
USD
BT124590-10G
BT124590-250G
BT124590-50G
Sodium diatrizoate hydrate ≥98%
Sodium diatrizoate hydrate
CAS: 737-31-5
Powder; Molecular Formula: C11H8I3N2NaO4·xH2O; MW: 635.90 (anhydrous)
Light Sensitive
- Powder
- Light Sensitive
Formula:
C₁₁H₈I₃N₂NaO₄·xH₂O |
CAS Number:
737-31-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...