5-Bromo-2-methox-3-methylypyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 5-Bromo-2-methoxy-3-methylpyridine

124580-1G 124580-5G
BT124580-1GEA 136 USD
BT124580-1G BT124580-5G
5-Bromo-2-methox-3-methylypyridine ≥97%
5-Bromo-2-methox-3-methylypyridine

CAS: 760207-87-2; MDL No: MFCD07375124 Liquid; Molecular Formula: C7H8BrNO; MW: 202.05


  • Liquid

Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD07375124
CAS Number: 760207-87-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR