6-Bromopicolinic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid

124555-1G 124555-5G
BT124555-1GEA 53 USD
BT124555-1G BT124555-5G
6-Bromopicolinic acid ≥97%
6-Bromopicolinic acid

CAS: 21190-87-4; MDL No: MFCD00093197 Crystalline; Molecular Formula: C6H3BrNO2; MW: 202.01 Melting Point: 192-194°


  • Crystalline

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 192…194 °C
MDL Number: MFCD00093197
CAS Number: 21190-87-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR