11-Bromo-1-undecene ≥96%

Supplier: BeanTown Chemical

124520-25G 124520-5G
BT124520-25GEA 194.2 USD
BT124520-25G BT124520-5G
11-Bromo-1-undecene ≥96%
11-Bromo-1-undecene

CAS: 7766-50-9; MDL No: MFCD00040825 Liquid; Linear Formula: Br(CH2)9CH=CH2; Molecular Formula: C11H21Br; MW: 233.19 Boiling Point: 149-150°/35 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.063; Refractive Index: 1.468


  • Refractive index: 1.468
  • Liquid

Formula: C₁₁H₂₁Br
MW: 233.19 g/mol
Boiling Pt: 149…150 °C/35 mmHg
Density: 1.063 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00040825
CAS Number: 7766-50-9

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR