L-serine methyl ester hydrochloride ≥98%
Supplier: BeanTown Chemical
CAS: 5680-80-8; EC No: 227-140-7; MDL No: MFCD00063680
Crystalline; Molecular Formula: C4H9NO3·HCl; MW: 155.58
Melting Point: 163° (decomposes)
Optical Rotation: [α]20/D +3.4°, c = 4 in methanol
Moisture Sensitive
- Optical rotation: [α]20/D +3.4°, c = 4 in methanol
- Moisture Sensitive
- Crystalline
Formula:
C₄H₉NO₃ Melting Pt: 163 °C (dec.) |
MDL Number:
MFCD00063680 CAS Number: 5680-80-8 EINECS: 227-140-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...