L(+)-Glutamine ≥99%
Supplier: BeanTown Chemical
Synonyms:
Q
, (S)-2,5-Diamino-5-oxopentanoic acid
, L-Glutamine
, Levoglutamide
, (S)-(+)-Glutamine
, Gln
, H-Gln-OH
124505-50G
124505-1KG
124505-250G
BT124505-1KGEA
346.92
USD
BT124505-1KG
BT124505-50G
BT124505-250G
L(+)-Glutamine ≥99%
L(+)-Glutamine
- RTECS: MA2275100
- Powder
Formula:
C₅H₁₀N₂O₃ MW: 146.15 g/mol Melting Pt: 185 °C (dec.) |
MDL Number:
MFCD00008044 CAS Number: 56-85-9 EINECS: 200-292-1 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...