(R)-(-)-4-Phenyl-2-oxazolidinone ≥98%
Supplier: BeanTown Chemical
Synonyms:
(R)-4-Phenyloxazolidin-2-one
124470-1G
124470-5G
BT124470-1GEA
72
USD
BT124470-1G
BT124470-5G
(R)-(-)-4-Phenyl-2-oxazolidinone ≥98%
(R)-(-)-4-Phenyl-2-oxazolidinone
CAS: 90319-52-1; MDL No: MFCD00192393
Crystalline; Molecular Formula: C9H9NO2 ; MW: 163.17
Melting Point: 131-133°
Optical Rotation: [α]25/D −48°, c = 2 in chloroform
- Optical rotation: [α]25/D −48°, c = 2 in chloroform
- Crystalline
Formula:
C₉H₉NO₂ MW: 163.18 g/mol Melting Pt: 131…133 °C |
MDL Number:
MFCD00192393 CAS Number: 90319-52-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...