Malonic acid ≥99.5%
Supplier: BeanTown Chemical
124445-100G
124445-2.5KG
124445-500G
BT124445-100GEA
43.83
USD
BT124445-100G
BT124445-2.5KG
BT124445-500G
Malonic acid ≥99.5%
Malonic acid
CAS: 141-82-2; EC No: 205-503-0; MDL No: MFCD00002707; RTECS: OO8175000
Powder; Linear Formula: CH2(COOH)2; MW: 104.06
Melting Point: 132-135° (decomposes); Flash point: 157°C (314°F)
Density (g/mL): 1.6
- RTECS: OO8175000
- Powder
Formula:
C₃H₄O₄ Melting Pt: 132…135 °C (dec.) Density: 1.6 g/mL Flash Pt: 157 °C (314 °F) |
MDL Number:
MFCD00002707 CAS Number: 141-82-2 EINECS: 205-503-0 |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...